Learn More
Grainger G3262 Level Gauge HDPE D 90in
Catalog No. 502764607
Encompass_Preferred
The Krueger Level Gauge For 90 in Container is a mechanical level gauge designed for use with 90 inch tall industrial containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and is adjustable to fit containers from 88 to 92 inches tall. It can handle temperatures up to 180°F and pressures up to 100 PSI. This heavy duty mechanical gauge provides simple, reliable level indication for industrial applications.
Catalog No. 50-276-4607 Supplier Grainger Supplier No. 16N274
The Krueger Level Gauge For 90 in Container is a mechanical level gauge designed for use with 90 inch tall industrial containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and is adjustable to fit containers from 88 to 92 inches tall. It can handle temperatures up to 180°F and pressures up to 100 PSI. This heavy duty mechanical gauge provides simple, reliable level indication for industrial applications.