Learn More
Grainger G3262 Level Gauge HDPE D 92in
Catalog No. 502764623
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include corrosion resistance, chemical compatibility, accuracy within 1/8", and visibility in daylight. They mount to the top or side of drums and tanks from 15 gallons up to 330 gallons. Multiple length options are available from 6 inches to 78 inches. These simple mechanical gauges provide an economical way to monitor liquid levels.
Catalog No. 50-276-4623 Supplier Grainger Supplier No. 16N276
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include corrosion resistance, chemical compatibility, accuracy within 1/8", and visibility in daylight. They mount to the top or side of drums and tanks from 15 gallons up to 330 gallons. Multiple length options are available from 6 inches to 78 inches. These simple mechanical gauges provide an economical way to monitor liquid levels.