Learn More
Grainger G3262 Level Gauge HDPE D 93in
Catalog No. 502764608
Encompass_Preferred
The Grainger liquid level gauges for drums and tanks are designed for use in 93 inch deep containers. They feature polypropylene and stainless steel construction with float and cage assemblies to provide continuous monitoring of liquid levels. These gauges have maximum liquid temperatures from 200-500°F depending on the model. They are available with standard or hazardous location approvals and have dial, digital, or magnetic readout options. Overall these gauges provide an accurate and durable solution for monitoring liquid levels in large volume tanks and drums.
Catalog No. 50-276-4608 Supplier Grainger Supplier No. 16N277
Add to Cart
Add to Cart
The Grainger liquid level gauges for drums and tanks are designed for use in 93 inch deep containers. They feature polypropylene and stainless steel construction with float and cage assemblies to provide continuous monitoring of liquid levels. These gauges have maximum liquid temperatures from 200-500°F depending on the model. They are available with standard or hazardous location approvals and have dial, digital, or magnetic readout options. Overall these gauges provide an accurate and durable solution for monitoring liquid levels in large volume tanks and drums.