Learn More
Grainger G3262 Level Gauge HDPE D 94in
Catalog No. 502764125
Encompass_Preferred
The Krueger Level Gauge For 94 in Container is a mechanical level gauge designed for use with 94 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and mounts to the top of the container with a 2 inch NPT connection. This heavy duty mechanical gauge provides accurate level indication without any power requirements. Made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-4125 Supplier Grainger Supplier No. 16N278
The Krueger Level Gauge For 94 in Container is a mechanical level gauge designed for use with 94 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and mounts to the top of the container with a 2 inch NPT connection. This heavy duty mechanical gauge provides accurate level indication without any power requirements. Made in the USA by Krueger Sentry Gauge.