Learn More
Grainger G3262 Level Gauge HDPE D 95in
Catalog No. 502764555
Encompass_Preferred
The Krueger Level Gauge for 95-gallon containers is a mechanical gauge with a stainless steel body and polypropylene float for chemical resistance. It shows the container level on an external scale in 1-gallon increments up to 95 gallons, allowing easy monitoring. The gauge threads into a standard 2-inch container opening, with a compression fitting to prevent leaks. This float gauge offers a simple, economical way to monitor levels in 95-gallon drums or tanks.
Catalog No. 50-276-4555 Supplier Grainger Supplier No. 16N279
The Krueger Level Gauge for 95-gallon containers is a mechanical gauge with a stainless steel body and polypropylene float for chemical resistance. It shows the container level on an external scale in 1-gallon increments up to 95 gallons, allowing easy monitoring. The gauge threads into a standard 2-inch container opening, with a compression fitting to prevent leaks. This float gauge offers a simple, economical way to monitor levels in 95-gallon drums or tanks.