Learn More
Grainger G3262 Level Gauge HDPE D 97in
Catalog No. 502764086
Encompass_Preferred
The Krueger Level Gauge For 97 in Container is a mechanical level gauge designed for use with 97 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and mounts to the top of the container with a 2 inch NPT connection. This heavy duty mechanical gauge provides accurate level indication without any power requirements.
Catalog No. 50-276-4086 Supplier Grainger Supplier No. 16N280
The Krueger Level Gauge For 97 in Container is a mechanical level gauge designed for use with 97 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 6 inch scale and mounts to the top of the container with a 2 inch NPT connection. This heavy duty mechanical gauge provides accurate level indication without any power requirements.