Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 97in
Catalog No. 502765866
Encompass_Preferred
The Krueger Level Gauge For 97-in Container is a mechanical level gauge designed for use with 97-inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch scale and indicates liquid level from 0 to 97 inches. This mechanical gauge requires no power and is ideal for visual monitoring of liquid levels in chemical tanks and vessels. With its durable construction, the Krueger level gauge provides reliable liquid level indication for industrial applications.
Catalog No. 50-276-5866 Supplier Grainger Supplier No. 16N573
The Krueger Level Gauge For 97-in Container is a mechanical level gauge designed for use with 97-inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch scale and indicates liquid level from 0 to 97 inches. This mechanical gauge requires no power and is ideal for visual monitoring of liquid levels in chemical tanks and vessels. With its durable construction, the Krueger level gauge provides reliable liquid level indication for industrial applications.