Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 98in
Catalog No. 502765867
Encompass_Preferred
The Krueger 16N574 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and other pressurized vessels to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and brass 1/4 inch NPT process connections. It is made in the USA and is rated for pressures up to 250 PSI at temperatures up to 750°F. The chamber length is 9 inches. This is a reliable and durable magnetic level gauge for indicating liquid level in high pressure, high temperature applications.
Catalog No. 50-276-5867 Supplier Grainger Supplier No. 16N574
The Krueger 16N574 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and other pressurized vessels to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and brass 1/4 inch NPT process connections. It is made in the USA and is rated for pressures up to 250 PSI at temperatures up to 750°F. The chamber length is 9 inches. This is a reliable and durable magnetic level gauge for indicating liquid level in high pressure, high temperature applications.