Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 86in
Catalog No. 502766042
Encompass_Preferred
The Krueger 16N563 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and process tanks to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and an aluminum case. It is made in the USA and is adjustable to fit tank connections from 5-3/4 inches to 7 inches. The level gauge comes with Teflon packing nuts and gaskets. It can withstand temperatures up to 750°F and pressures up to 500 psi. The magnetic level gauge provides a visual indication of liquid level in a closed tank or boiler.
Catalog No. 50-276-6042 Supplier Grainger Supplier No. 16N563
The Krueger 16N563 is a magnetic level gauge designed for use with steam boilers, hot water heating systems, and process tanks to indicate liquid level. This level gauge has a 316 stainless steel chamber with mica viewing windows and an aluminum case. It is made in the USA and is adjustable to fit tank connections from 5-3/4 inches to 7 inches. The level gauge comes with Teflon packing nuts and gaskets. It can withstand temperatures up to 750°F and pressures up to 500 psi. The magnetic level gauge provides a visual indication of liquid level in a closed tank or boiler.