Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 66in
Catalog No. 502766043
Encompass_Preferred
The Krueger 16N545 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale graduated from 0 to 100% with 2% increments. The gauge has a stainless steel float which moves vertically within a stainless steel guide tube as liquid level changes. It includes top and bottom stainless steel flanges for mounting. This mechanical float-type gauge requires no power and is suitable for use with water-based chemicals and petroleum products. With its rugged stainless construction, the Krueger 16N545 provides a simple, economical way to monitor liquid level.
Catalog No. 50-276-6043 Supplier Grainger Supplier No. 16N545
The Krueger 16N545 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch scale graduated from 0 to 100% with 2% increments. The gauge has a stainless steel float which moves vertically within a stainless steel guide tube as liquid level changes. It includes top and bottom stainless steel flanges for mounting. This mechanical float-type gauge requires no power and is suitable for use with water-based chemicals and petroleum products. With its rugged stainless construction, the Krueger 16N545 provides a simple, economical way to monitor liquid level.