Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 65in
Catalog No. 502767122
Encompass_Preferred
The Krueger 16N544 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be used for a wide variety of liquids. The gauge has a dual scale that reads from 0 to 100 percent in 2 percent increments on the front and 0 to 10 inches on the back. It has a maximum temperature rating of 250 degrees Fahrenheit. With its rugged industrial design, the Krueger 16N544 provides accurate liquid level indication for process monitoring.
Catalog No. 50-276-7122 Supplier Grainger Supplier No. 16N544
The Krueger 16N544 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be used for a wide variety of liquids. The gauge has a dual scale that reads from 0 to 100 percent in 2 percent increments on the front and 0 to 10 inches on the back. It has a maximum temperature rating of 250 degrees Fahrenheit. With its rugged industrial design, the Krueger 16N544 provides accurate liquid level indication for process monitoring.