Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 68in
Numéro de catalogue. 502767127
Encompass_Preferred
The Krueger 16N547 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. It has a 3" dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid level in tanks or vessels. It can be mounted to the top or side of a tank. This industrial liquid level gauge is made in the USA by Krueger Sentry Gauge.
Numéro de catalogue. 50-276-7127 Fournisseur Grainger Code fournisseur 16N547
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Krueger 16N547 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. It has a 3" dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid level in tanks or vessels. It can be mounted to the top or side of a tank. This industrial liquid level gauge is made in the USA by Krueger Sentry Gauge.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.