Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 79in
Catalog No. 502764955
Encompass_Preferred
The Krueger Level Gauge For 79 in Container is a mechanical level gauge designed for use with 79 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 10 inch scale marked in 1/4 inch increments to provide an accurate reading. This heavy duty industrial gauge can withstand temperatures up to 180°F and has a maximum pressure rating of 100 PSI. The gauge threads directly into a standard 2 inch opening and is made in the USA.
Catalog No. 50-276-4955 Supplier Grainger Supplier No. 16N557
The Krueger Level Gauge For 79 in Container is a mechanical level gauge designed for use with 79 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a 10 inch scale marked in 1/4 inch increments to provide an accurate reading. This heavy duty industrial gauge can withstand temperatures up to 180°F and has a maximum pressure rating of 100 PSI. The gauge threads directly into a standard 2 inch opening and is made in the USA.