Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 103in
Catalog No. 502765021
Encompass_Preferred
The Krueger Level Gauge For 102 in Container is a float-operated mechanical level gauge designed for use in 102 inch high containers. It features a hermetically sealed gauge body and float for use with water-based chemicals. The gauge indicates liquid level on a clearly marked external scale in 1/2 inch increments. It has a stainless steel float and polymer coated steel casing. The gauge installs easily through a tank top fitting and is made in the USA. This rugged and economical mechanical gauge is a reliable way to monitor liquid level in large chemical tanks.
Catalog No. 50-276-5021 Supplier Grainger Supplier No. 16N579
The Krueger Level Gauge For 102 in Container is a float-operated mechanical level gauge designed for use in 102 inch high containers. It features a hermetically sealed gauge body and float for use with water-based chemicals. The gauge indicates liquid level on a clearly marked external scale in 1/2 inch increments. It has a stainless steel float and polymer coated steel casing. The gauge installs easily through a tank top fitting and is made in the USA. This rugged and economical mechanical gauge is a reliable way to monitor liquid level in large chemical tanks.