Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 32in
Catalog No. 502765667
Encompass_Preferred
The Grainger 16N519 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with 3/8-inch drive ratchets, handles, extensions, etc. It measures 1-1/2 inches long and weighs 0.8 ounces. The 16N519 has a tensile strength of 100,000 PSI.
Catalog No. 50-276-5667 Supplier Grainger Supplier No. 16N519
The Grainger 16N519 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with 3/8-inch drive ratchets, handles, extensions, etc. It measures 1-1/2 inches long and weighs 0.8 ounces. The 16N519 has a tensile strength of 100,000 PSI.