Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 14in
Catalog No. 502765854
Encompass_Preferred
The Krueger Level Gauge For 18 in Container is a liquid level indicator designed for 18 inch diameter tanks. It features a polypropylene float that moves up and down a stainless steel guide rod as liquid level changes. The gauge has a maximum liquid height of 48 inches and includes a scale to easily read the liquid level. This industrial gauge helps monitor liquid levels in processing and storage tanks. It has a corrosion resistant design suitable for use with water, chemicals, petroleum products and more. The Krueger level gauge provides an economical way to monitor tank fill levels.
Catalog No. 50-276-5854 Supplier Grainger Supplier No. 16N503
The Krueger Level Gauge For 18 in Container is a liquid level indicator designed for 18 inch diameter tanks. It features a polypropylene float that moves up and down a stainless steel guide rod as liquid level changes. The gauge has a maximum liquid height of 48 inches and includes a scale to easily read the liquid level. This industrial gauge helps monitor liquid levels in processing and storage tanks. It has a corrosion resistant design suitable for use with water, chemicals, petroleum products and more. The Krueger level gauge provides an economical way to monitor tank fill levels.