Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 33in
Catalog No. 502766198
Encompass_Preferred
The Krueger Level Gauge For 102 in Container is a liquid level gauge designed for 102 inch tall containers. It features a polypropylene float that moves up and down a stainless steel tape to indicate liquid level. The gauge has a maximum reading of 102 inches and includes markings every 1/4 inch for precise measurement. This industrial gauge helps monitor liquid levels in large tanks and containers. With its durable construction, it provides reliable liquid level indication for applications such as chemical processing, waste water treatment, and more. The gauge mounts to the top of containers with a 3/4 inch NPT connection.
Catalog No. 50-276-6198 Supplier Grainger Supplier No. 16N520
The Krueger Level Gauge For 102 in Container is a liquid level gauge designed for 102 inch tall containers. It features a polypropylene float that moves up and down a stainless steel tape to indicate liquid level. The gauge has a maximum reading of 102 inches and includes markings every 1/4 inch for precise measurement. This industrial gauge helps monitor liquid levels in large tanks and containers. With its durable construction, it provides reliable liquid level indication for applications such as chemical processing, waste water treatment, and more. The gauge mounts to the top of containers with a 3/4 inch NPT connection.