Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 27in
Catalog No. 502767571
Encompass_Preferred
The Krueger 16N515 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a scale range of 0-60 inches. The polypropylene case is chemical resistant with a maximum temperature rating of 200°F. This mechanical gauge requires no electricity and indicates the liquid level on an external scale. The 16N515 has 3/4 inch NPT connections and is made in the USA. It can be used for a variety of liquids including water, oils, and chemicals. This direct reading gauge provides simple visual monitoring of tank levels.
Catalog No. 50-276-7571 Supplier Grainger Supplier No. 16N515
The Krueger 16N515 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a scale range of 0-60 inches. The polypropylene case is chemical resistant with a maximum temperature rating of 200°F. This mechanical gauge requires no electricity and indicates the liquid level on an external scale. The 16N515 has 3/4 inch NPT connections and is made in the USA. It can be used for a variety of liquids including water, oils, and chemicals. This direct reading gauge provides simple visual monitoring of tank levels.