Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 57in
Catalog No. 502767572
Encompass_Preferred
The Krueger Level Gauge For 57 in Container is a mechanical level gauge designed for use with 57 inch diameter containers. It features a stainless steel body and float construction with a polypropylene indicator that provides an easy visual reference for determining liquid level. This gauge has a maximum liquid level reading of 48 inches and can withstand operating temperatures between -40 and 250 degrees Fahrenheit. With its durable materials, wide measurement range, and easy-to-read display, this mechanical gauge is well-suited for monitoring liquid levels in large containers.
Catalog No. 50-276-7572 Supplier Grainger Supplier No. 16N538
The Krueger Level Gauge For 57 in Container is a mechanical level gauge designed for use with 57 inch diameter containers. It features a stainless steel body and float construction with a polypropylene indicator that provides an easy visual reference for determining liquid level. This gauge has a maximum liquid level reading of 48 inches and can withstand operating temperatures between -40 and 250 degrees Fahrenheit. With its durable materials, wide measurement range, and easy-to-read display, this mechanical gauge is well-suited for monitoring liquid levels in large containers.