Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 92in
Catalog No. 502767621
Encompass_Preferred
The Grainger 16N569 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing electric motor. It has a 1/2" diameter shaft and measures approximately 9-1/2" long by 5-3/4" wide. This motor can be used for a variety of industrial applications and is totally enclosed and fan cooled for protection against dirt, dust and splashing water. It requires a minimum 12 gauge wire and 30 amp circuit breaker and operates on single phase 115/230V power. This electric motor is CSA certified and backed by a 1 year warranty from Grainger.
Catalog No. 50-276-7621 Supplier Grainger Supplier No. 16N569
The Grainger 16N569 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing electric motor. It has a 1/2" diameter shaft and measures approximately 9-1/2" long by 5-3/4" wide. This motor can be used for a variety of industrial applications and is totally enclosed and fan cooled for protection against dirt, dust and splashing water. It requires a minimum 12 gauge wire and 30 amp circuit breaker and operates on single phase 115/230V power. This electric motor is CSA certified and backed by a 1 year warranty from Grainger.