Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 37in
Catalog No. 502767751
Encompass_Preferred
The Krueger Level Gauge For 18 in Container is a liquid level indicator designed for 18 inch diameter tanks. It features a polypropylene float that moves up and down a stainless steel guide rod as liquid level changes. The gauge has a maximum liquid level indication of 15 inches and includes adhesive for mounting. This mechanical float gauge provides at-a-glance monitoring of liquid level in a variety of applications. It is suitable for use with water, oil and other non-corrosive liquids. The gauge measures 3 inches wide by 16 inches high.
Catalog No. 50-276-7751 Supplier Grainger Supplier No. 16N523
The Krueger Level Gauge For 18 in Container is a liquid level indicator designed for 18 inch diameter tanks. It features a polypropylene float that moves up and down a stainless steel guide rod as liquid level changes. The gauge has a maximum liquid level indication of 15 inches and includes adhesive for mounting. This mechanical float gauge provides at-a-glance monitoring of liquid level in a variety of applications. It is suitable for use with water, oil and other non-corrosive liquids. The gauge measures 3 inches wide by 16 inches high.