Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 101in
Catalog No. 502768314
Encompass_Preferred
The Krueger 16N577 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual indication of fluid level. The gauge body is made of steel and mounts via a 1/2" NPT thread. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The gauge measures 5 inches long with a 2 inch diameter. Overall, this is a heavy duty mechanical fluid level gauge for industrial equipment that provides an easy visual reference for proper oil level.
Catalog No. 50-276-8314 Supplier Grainger Supplier No. 16N577
The Krueger 16N577 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual indication of fluid level. The gauge body is made of steel and mounts via a 1/2" NPT thread. It has a maximum pressure rating of 1000 PSI and temperature rating of 250°F. The gauge measures 5 inches long with a 2 inch diameter. Overall, this is a heavy duty mechanical fluid level gauge for industrial equipment that provides an easy visual reference for proper oil level.