Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 50in
Catalog No. 502768524
Encompass_Preferred
The Krueger 16N531 Level Gauge is a liquid level indicator designed for use in industrial tanks and vessels. It features a polypropylene float that moves up and down a 316 stainless steel stem in response to liquid level changes. The indicator dial shows the liquid level and can be customized with gallonage markers. This mechanical gauge has a maximum temperature rating of 180°F and maximum pressure rating of 100 PSI. It has 3/4" NPT connections and is available in lengths from 7" to 48". The Krueger 16N531 provides a simple, economical way to monitor liquid levels.
Catalog No. 50-276-8524 Supplier Grainger Supplier No. 16N531
The Krueger 16N531 Level Gauge is a liquid level indicator designed for use in industrial tanks and vessels. It features a polypropylene float that moves up and down a 316 stainless steel stem in response to liquid level changes. The indicator dial shows the liquid level and can be customized with gallonage markers. This mechanical gauge has a maximum temperature rating of 180°F and maximum pressure rating of 100 PSI. It has 3/4" NPT connections and is available in lengths from 7" to 48". The Krueger 16N531 provides a simple, economical way to monitor liquid levels.