Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 51in
Catalog No. 502768525
Encompass_Preferred
The Krueger 16N532 is a liquid level gauge designed for use with hydraulic reservoirs and gearboxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in both inches and millimeters with a maximum range of 0-8 inches/0-200mm. It uses a bi-color indicator to clearly show the current fluid level. This mechanical float gauge can withstand operating temperatures between -40 and 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection. The 16N532 level gauge is ideal for measuring oil level in industrial equipment.
Catalog No. 50-276-8525 Supplier Grainger Supplier No. 16N532
The Krueger 16N532 is a liquid level gauge designed for use with hydraulic reservoirs and gearboxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in both inches and millimeters with a maximum range of 0-8 inches/0-200mm. It uses a bi-color indicator to clearly show the current fluid level. This mechanical float gauge can withstand operating temperatures between -40 and 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection. The 16N532 level gauge is ideal for measuring oil level in industrial equipment.