Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 54in
Catalog No. 502768526
Encompass_Preferred
The Krueger 16N535 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a clearly marked scale and has a maximum working pressure of 150 PSI. This rugged and durable gauge allows operators to monitor liquid levels in tanks or vessels visually. With proper installation, it provides accurate liquid level readings to help optimize processes and prevent overfills or run-dry conditions. The 16N535 is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-8526 Supplier Grainger Supplier No. 16N535
The Krueger 16N535 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a clearly marked scale and has a maximum working pressure of 150 PSI. This rugged and durable gauge allows operators to monitor liquid levels in tanks or vessels visually. With proper installation, it provides accurate liquid level readings to help optimize processes and prevent overfills or run-dry conditions. The 16N535 is made in the USA by Krueger Sentry Gauge.