Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 56in
Catalog No. 502768527
Encompass_Preferred
The Krueger 16N537 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color float to indicate level - green for normal level and red when the level drops below 3.5 inches. The gauge can withstand temperatures from -40 to 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection and comes with a locknut for easy installation on hydraulic reservoirs. The gauge allows operators to monitor fluid levels at a glance to ensure proper lubrication.
Catalog No. 50-276-8527 Supplier Grainger Supplier No. 16N537
The Krueger 16N537 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that reads in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color float to indicate level - green for normal level and red when the level drops below 3.5 inches. The gauge can withstand temperatures from -40 to 257 degrees Fahrenheit. It has a 1/2 inch NPT thread connection and comes with a locknut for easy installation on hydraulic reservoirs. The gauge allows operators to monitor fluid levels at a glance to ensure proper lubrication.