Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 39in
Catalog No. 502768528
Encompass_Preferred
The Krueger 16N524 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be used for hazardous locations. The gauge has a dual scale that reads from 0-100% and 0-10 inches. It can withstand operating temperatures from -40 to 250°F. The gauge comes with a 3/4 inch mounting connection and requires a 1/2 inch NPT tank connection. It has an accuracy of ±5%.
Catalog No. 50-276-8528 Supplier Grainger Supplier No. 16N524
The Krueger 16N524 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be used for hazardous locations. The gauge has a dual scale that reads from 0-100% and 0-10 inches. It can withstand operating temperatures from -40 to 250°F. The gauge comes with a 3/4 inch mounting connection and requires a 1/2 inch NPT tank connection. It has an accuracy of ±5%.