Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 87in
Catalog No. 502768601
Encompass_Preferred
The Krueger 16N564 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2" diameter. The gauge meets ASME B31.1 and B31.3 requirements. With a maximum pressure rating of 30 psi @ 100°F, this Krueger level gauge provides an economical solution for monitoring liquid levels in process tanks.
Catalog No. 50-276-8601 Supplier Grainger Supplier No. 16N564
The Krueger 16N564 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2" diameter. The gauge meets ASME B31.1 and B31.3 requirements. With a maximum pressure rating of 30 psi @ 100°F, this Krueger level gauge provides an economical solution for monitoring liquid levels in process tanks.