Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 88in
Catalog No. 502768602
Encompass_Preferred
The Krueger 16N565 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2” and is rated for pressures up to 500 psi at temperatures up to 450°F. The Krueger 16N565 provides continuous monitoring of tank levels for various industrial applications.
Catalog No. 50-276-8602 Supplier Grainger Supplier No. 16N565
Add to Cart
Add to Cart
The Krueger 16N565 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid levels. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2” and is rated for pressures up to 500 psi at temperatures up to 450°F. The Krueger 16N565 provides continuous monitoring of tank levels for various industrial applications.