Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 17in
Catalog No. 502768604
Encompass_Preferred
The Krueger Level Gauge For 18 in Container is a liquid level gauge designed for 18 inch diameter containers. It features a stainless steel construction with a polypropylene float that provides chemical resistance. The gauge has a maximum reading of 16 inches and includes rear mounting studs for easy installation. This industrial gauge helps monitor liquid levels in containers up to 18 inches in diameter. With its durable stainless steel and polypropylene design, it offers an accurate and reliable solution for monitoring container fluid levels.
Catalog No. 50-276-8604 Supplier Grainger Supplier No. 16N506
The Krueger Level Gauge For 18 in Container is a liquid level gauge designed for 18 inch diameter containers. It features a stainless steel construction with a polypropylene float that provides chemical resistance. The gauge has a maximum reading of 16 inches and includes rear mounting studs for easy installation. This industrial gauge helps monitor liquid levels in containers up to 18 inches in diameter. With its durable stainless steel and polypropylene design, it offers an accurate and reliable solution for monitoring container fluid levels.