Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 26in
Catalog No. 502768606
Encompass_Preferred
The Krueger 16N514 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that measures in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color indicator to clearly show the current fluid level. This mechanical float gauge can withstand operating temperatures from -40 to 250°F. It has a 1/2 inch NPT thread connection. The 16N514 provides at-a-glance monitoring of fluid levels.
Catalog No. 50-276-8606 Supplier Grainger Supplier No. 16N514
The Krueger 16N514 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a polycarbonate lens and aluminum housing for durability. The gauge has a dual scale that measures in inches and centimeters with a max level of 8 inches or 20 cm. It uses a bi-color indicator to clearly show the current fluid level. This mechanical float gauge can withstand operating temperatures from -40 to 250°F. It has a 1/2 inch NPT thread connection. The 16N514 provides at-a-glance monitoring of fluid levels.