Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 69in
Catalog No. 502766494
Encompass_Preferred
The Krueger Level Gauge For 18 in Container is a liquid level indicator designed for 18 inch diameter tanks. It features a clear polycarbonate gauge body with a stainless steel float and polypropylene float stop. The gauge indicates liquid level on a easy-to-read scale in 1/8 inch increments. It has a maximum temperature rating of 180°F and fits standard 2 inch couplings. This rugged and economical gauge helps monitor liquid levels in industrial tanks.
Catalog No. 50-276-6494 Supplier Grainger Supplier No. 16N548
The Krueger Level Gauge For 18 in Container is a liquid level indicator designed for 18 inch diameter tanks. It features a clear polycarbonate gauge body with a stainless steel float and polypropylene float stop. The gauge indicates liquid level on a easy-to-read scale in 1/8 inch increments. It has a maximum temperature rating of 180°F and fits standard 2 inch couplings. This rugged and economical gauge helps monitor liquid levels in industrial tanks.