Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 43in
Catalog No. 502767196
Encompass_Preferred
The Krueger 16N527 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 PSI. It has a 3 inch dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid level in tanks or vessels. It can be mounted to the top or side of a tank. This economical gauge is ideal for monitoring level in water, oils, chemicals, and more. The 16N527 helps optimize tank utilization and prevent overfills.
Catalog No. 50-276-7196 Supplier Grainger Supplier No. 16N527
The Krueger 16N527 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 PSI. It has a 3 inch dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid level in tanks or vessels. It can be mounted to the top or side of a tank. This economical gauge is ideal for monitoring level in water, oils, chemicals, and more. The 16N527 helps optimize tank utilization and prevent overfills.