Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 59in
Catalog No. 502767398
Encompass_Preferred
The Krueger 102-inch Level Gauge is a liquid level indicator designed for 102-inch containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to show the liquid level. The gauge has a hermetically sealed design to prevent leakage and is adjustable to fit different container heights. It has a maximum temperature rating of 250°F and includes mounting hardware for installation. This industrial gauge helps monitor liquid levels in large tanks and vessels.
Catalog No. 50-276-7398 Supplier Grainger Supplier No. 16N540
The Krueger 102-inch Level Gauge is a liquid level indicator designed for 102-inch containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to show the liquid level. The gauge has a hermetically sealed design to prevent leakage and is adjustable to fit different container heights. It has a maximum temperature rating of 250°F and includes mounting hardware for installation. This industrial gauge helps monitor liquid levels in large tanks and vessels.