Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 91in
Catalog No. 502765568
Encompass_Preferred
The Krueger 16N568 is a liquid level gauge designed for use in industrial tanks and vessels to monitor fluid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be mounted outside the tank. It has a dual scale that reads from 0-100% in 2% increments on the front and 0-10 inches on the back. The gauge comes ready to install with a brass shutoff valve, process connection, and fittings. It is made in the USA by Krueger Sentry Gauge. This level gauge provides an accurate visual monitoring solution for industrial fluid tanks.
Catalog No. 50-276-5568 Supplier Grainger Supplier No. 16N568
The Krueger 16N568 is a liquid level gauge designed for use in industrial tanks and vessels to monitor fluid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed magnetic coupling that allows the gauge to be mounted outside the tank. It has a dual scale that reads from 0-100% in 2% increments on the front and 0-10 inches on the back. The gauge comes ready to install with a brass shutoff valve, process connection, and fittings. It is made in the USA by Krueger Sentry Gauge. This level gauge provides an accurate visual monitoring solution for industrial fluid tanks.