Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 82in
Catalog No. 502765504
Encompass_Preferred
The Krueger Level Gauge For 82 in Container is a mechanical level gauge designed for use with 82 inch high containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a scale range from 0 to 82 inches and is suitable for use with water, wastewater, chemicals, petroleum products and food processing liquids. This mechanical gauge requires no power and is built for durability with stainless steel and polypropylene wetted parts. It can withstand operating temperatures up to 180°F. The Krueger model number for this 82 inch level gauge is 16N560.
Catalog No. 50-276-5504 Supplier Grainger Supplier No. 16N560
The Krueger Level Gauge For 82 in Container is a mechanical level gauge designed for use with 82 inch high containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a scale range from 0 to 82 inches and is suitable for use with water, wastewater, chemicals, petroleum products and food processing liquids. This mechanical gauge requires no power and is built for durability with stainless steel and polypropylene wetted parts. It can withstand operating temperatures up to 180°F. The Krueger model number for this 82 inch level gauge is 16N560.