Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 63in
Catalog No. 502765506
Encompass_Preferred
The Krueger 16N543 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The dial is oil-filled to dampen fluctuation. Overall, the 16N543 provides simple, rugged liquid level indication.
Catalog No. 50-276-5506 Supplier Grainger Supplier No. 16N543
The Krueger 16N543 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The dial is oil-filled to dampen fluctuation. Overall, the 16N543 provides simple, rugged liquid level indication.