Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 111in
Catalog No. 502765589
Encompass_Preferred
The Krueger 16N586 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual display of the tank level through glass windows on the gauge. The gauge can withstand temperatures up to 500°F and pressures up to 500 psi. It has 3/4 inch NPT process connections and is designed for use with water, oils, and other non-coating media. This is an economical and easy-to-read liquid level indicator.
Catalog No. 50-276-5589 Supplier Grainger Supplier No. 16N586
The Krueger 16N586 is a magnetic level gauge that indicates liquid level in tanks and vessels. It has a 316 stainless steel chamber and Teflon float with a magnetic coupling that activates red and green indicator flags outside the chamber as the float rises and falls with the liquid level. This provides a visual display of the tank level through glass windows on the gauge. The gauge can withstand temperatures up to 500°F and pressures up to 500 psi. It has 3/4 inch NPT process connections and is designed for use with water, oils, and other non-coating media. This is an economical and easy-to-read liquid level indicator.