Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 83in
Catalog No. 502765811
Encompass_Preferred
The Krueger 16N561 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch clear acrylic tube with color coded polypropylene float. It has flanged top and bottom fittings for easy installation. The gauge has a maximum pressure rating of 100 PSI and temperature rating of 180 degrees Fahrenheit. It is compatible with water, oil and mild chemicals. The overall length is 27 inches. This economical gauge provides simple, visual monitoring of liquid level.
Catalog No. 50-276-5811 Supplier Grainger Supplier No. 16N561
The Krueger 16N561 is a liquid level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. This model features a 10 inch clear acrylic tube with color coded polypropylene float. It has flanged top and bottom fittings for easy installation. The gauge has a maximum pressure rating of 100 PSI and temperature rating of 180 degrees Fahrenheit. It is compatible with water, oil and mild chemicals. The overall length is 27 inches. This economical gauge provides simple, visual monitoring of liquid level.