Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 13in
Catalog No. 502765988
Encompass_Preferred
The Krueger 16N502 is a magnetic level gauge that provides continuous liquid level indication for steam boilers, hot water heating systems, and other pressurized vessels. This level gauge has a 316 stainless steel chamber with a float and follower magnet inside that rises and falls with the liquid level. The level is displayed on a clear polycarbonate front cover which has a scaled indicator to show the level. This gauge has a maximum pressure rating of 30 psi and temperature rating of 250°F. The chamber size is 2" diameter x 16" length. Made in the USA by Krueger.
Catalog No. 50-276-5988 Supplier Grainger Supplier No. 16N502
The Krueger 16N502 is a magnetic level gauge that provides continuous liquid level indication for steam boilers, hot water heating systems, and other pressurized vessels. This level gauge has a 316 stainless steel chamber with a float and follower magnet inside that rises and falls with the liquid level. The level is displayed on a clear polycarbonate front cover which has a scaled indicator to show the level. This gauge has a maximum pressure rating of 30 psi and temperature rating of 250°F. The chamber size is 2" diameter x 16" length. Made in the USA by Krueger.