Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 117in
Catalog No. 502766011
Encompass_Preferred
The Krueger Level Gauge For 117 in Container is a level gauge designed for 117 inch containers. It features a polypropylene float that provides level indication through a clear, shatterproof lens. This mechanical gauge has a lightweight aluminum alloy body and is adjustable to different dimension tanks. It has a maximum temperature rating of 180°F and a minimum specific gravity rating of 0.5. The overall dimensions are 15 in x 5 in x 2 in. This level gauge is simple to install and provides reliable fluid level readings.
Catalog No. 50-276-6011 Supplier Grainger Supplier No. 16N592
The Krueger Level Gauge For 117 in Container is a level gauge designed for 117 inch containers. It features a polypropylene float that provides level indication through a clear, shatterproof lens. This mechanical gauge has a lightweight aluminum alloy body and is adjustable to different dimension tanks. It has a maximum temperature rating of 180°F and a minimum specific gravity rating of 0.5. The overall dimensions are 15 in x 5 in x 2 in. This level gauge is simple to install and provides reliable fluid level readings.