Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 18in
Catalog No. 502766075
Encompass_Preferred
The Krueger Level Gauge For 18 in Container is a liquid level gauge designed for 18 inch diameter containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has a maximum liquid height of 15 inches. It can be used with a variety of liquids including water, oil, chemicals, and more. The gauge helps monitor liquid levels at a glance without opening the container. It installs easily by drilling a 1-1/8 inch hole in the container wall and tightening the bulkhead fitting.
Catalog No. 50-276-6075 Supplier Grainger Supplier No. 16N507
The Krueger Level Gauge For 18 in Container is a liquid level gauge designed for 18 inch diameter containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has a maximum liquid height of 15 inches. It can be used with a variety of liquids including water, oil, chemicals, and more. The gauge helps monitor liquid levels at a glance without opening the container. It installs easily by drilling a 1-1/8 inch hole in the container wall and tightening the bulkhead fitting.