Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 28in
Catalog No. 502766094
Encompass_Preferred
The Krueger 16N516 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to resist corrosion, with a polypropylene indicator and EPDM seals for chemical resistance. The gauge has a maximum pressure rating of 150 PSI and temperature rating of 250°F. It has a 4 inch dial with a dual scale calibrated in inches and centimeters for reading liquid level. This mechanical float gauge provides continuous level indication through a 270 degree arc in vertical or horizontal orientation. It can be panel, surface or pipe mounted. The 16N516 is ideal for monitoring liquid levels in tanks, day tanks, hydraulic reservoirs and process vessels.
Catalog No. 50-276-6094 Supplier Grainger Supplier No. 16N516
The Krueger 16N516 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to resist corrosion, with a polypropylene indicator and EPDM seals for chemical resistance. The gauge has a maximum pressure rating of 150 PSI and temperature rating of 250°F. It has a 4 inch dial with a dual scale calibrated in inches and centimeters for reading liquid level. This mechanical float gauge provides continuous level indication through a 270 degree arc in vertical or horizontal orientation. It can be panel, surface or pipe mounted. The 16N516 is ideal for monitoring liquid levels in tanks, day tanks, hydraulic reservoirs and process vessels.