Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 76in
Catalog No. 502766153
Encompass_Preferred
The Krueger 16N554 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 PSI. It has a 3 inch dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid levels in tanks or vessels. It can be mounted to the top or side of a tank. This industrial liquid level gauge is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-6153 Supplier Grainger Supplier No. 16N554
The Krueger 16N554 is a liquid level gauge designed for industrial applications. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 PSI. It has a 3 inch dial with a dual scale calibrated in inches and centimeters. The gauge provides continuous monitoring of liquid levels in tanks or vessels. It can be mounted to the top or side of a tank. This industrial liquid level gauge is made in the USA by Krueger Sentry Gauge.