Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 70in
Catalog No. 502766156
Encompass_Preferred
The Krueger 70-inch Level Gauge is a liquid level indicator designed for 70-inch containers. It features a polypropylene float that moves up and down a stainless steel tape to show the liquid level. The gauge has a maximum reading of 70 inches and includes markings every 1/8 inch for precise measurement. This direct reading gauge requires no power source and is ideal for use with water-based chemicals in industrial applications. With its corrosion resistant construction, the Krueger Level Gauge provides an accurate and reliable way to monitor liquid levels in large containers.
Catalog No. 50-276-6156 Supplier Grainger Supplier No. 16N549
The Krueger 70-inch Level Gauge is a liquid level indicator designed for 70-inch containers. It features a polypropylene float that moves up and down a stainless steel tape to show the liquid level. The gauge has a maximum reading of 70 inches and includes markings every 1/8 inch for precise measurement. This direct reading gauge requires no power source and is ideal for use with water-based chemicals in industrial applications. With its corrosion resistant construction, the Krueger Level Gauge provides an accurate and reliable way to monitor liquid levels in large containers.