Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 94in
Catalog No. 502768474
Encompass_Preferred
The Krueger 16N571 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The dial is oil-filled to dampen fluctuation. An adjustable pointer kit is included for calibration. The gauge has 1/2 inch NPT male thread connections. It requires a minimum liquid specific gravity of 0.75 for proper operation. This industrial liquid level gauge provides an analog display of tank level.
Catalog No. 50-276-8474 Supplier Grainger Supplier No. 16N571
The Krueger 16N571 is a liquid level gauge designed for industrial applications. It features a stainless steel body and float to withstand harsh environments. The gauge indicates liquid level on a 6 inch dial with a 270 degree scale and has a maximum pressure rating of 150 PSI. This mechanical gauge can be used on a variety of tanks and vessels to monitor liquid levels. The dial is oil-filled to dampen fluctuation. An adjustable pointer kit is included for calibration. The gauge has 1/2 inch NPT male thread connections. It requires a minimum liquid specific gravity of 0.75 for proper operation. This industrial liquid level gauge provides an analog display of tank level.