Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 16in
Catalog No. 502768536
Encompass_Preferred
The Krueger 16N505 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The stem lengths range from 12 inches to 60 inches. This rugged and economical gauge provides accurate monitoring of liquid levels in a variety of applications. With proper installation, it can help prevent tank overfills or running dry. The Krueger 16N505 is an affordable, durable solution for industrial liquid level indication.
Catalog No. 50-276-8536 Supplier Grainger Supplier No. 16N505
The Krueger 16N505 is a liquid level gauge designed for use in industrial tanks and vessels to monitor liquid levels. It features a stainless steel float and stem with a polypropylene indicator. The gauge has a maximum temperature rating of 250°F and a maximum pressure rating of 150 psi. The stem lengths range from 12 inches to 60 inches. This rugged and economical gauge provides accurate monitoring of liquid levels in a variety of applications. With proper installation, it can help prevent tank overfills or running dry. The Krueger 16N505 is an affordable, durable solution for industrial liquid level indication.