Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 53in
Catalog No. 502765724
Encompass_Preferred
The Krueger 16N534 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual monitoring of oil level. The gauge has a 1/2 inch NPT thread and is made from durable brass construction. It has a maximum pressure rating of 1000 PSI and temperature rating of 250 degrees Fahrenheit. The Krueger 16N534 provides a simple way to monitor fluid levels in industrial equipment.
Catalog No. 50-276-5724 Supplier Grainger Supplier No. 16N534
The Krueger 16N534 is a liquid level gauge designed for use with hydraulic reservoirs and gear boxes. It features a clear acrylic window with green and red zones for quick visual monitoring of oil level. The gauge has a 1/2 inch NPT thread and is made from durable brass construction. It has a maximum pressure rating of 1000 PSI and temperature rating of 250 degrees Fahrenheit. The Krueger 16N534 provides a simple way to monitor fluid levels in industrial equipment.