Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 35in
Catalog No. 502766235
Encompass_Preferred
The Krueger 16N522 is a liquid level gauge designed for use in industrial tanks and vessels to monitor fluid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid level. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2" diameter. The gauge meets ASME B31.1 and B31.3 requirements. With a maximum pressure rating of 30 psi @ 100°F, this level gauge provides an accurate visual monitoring solution for a range of liquid level measurement applications.
Catalog No. 50-276-6235 Supplier Grainger Supplier No. 16N522
The Krueger 16N522 is a liquid level gauge designed for use in industrial tanks and vessels to monitor fluid levels. This model features a 10 inch scale length in stainless steel construction with a hermetically sealed gauge glass for viewing fluid level. It has 316 stainless steel flanged process connections available in sizes from 1/2" to 2" diameter. The gauge meets ASME B31.1 and B31.3 requirements. With a maximum pressure rating of 30 psi @ 100°F, this level gauge provides an accurate visual monitoring solution for a range of liquid level measurement applications.