Learn More
Grainger G3268 Level Gauge 304 Stainless Steel D 45in
Catalog No. 502766236
Encompass_Preferred
The Krueger Level Gauge For 102 in Container is a level gauge designed for use with 102 inch tall containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has gallon increments marked on the tube for easy monitoring of liquid level. This industrial gauge helps ensure proper liquid fill levels in large containers up to 102 inches tall. The gauge mounts to the top of the container and requires a 1-1/4 inch NPT opening. It has a maximum temperature rating of 150°F.
Catalog No. 50-276-6236 Supplier Grainger Supplier No. 16N528
The Krueger Level Gauge For 102 in Container is a level gauge designed for use with 102 inch tall containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has gallon increments marked on the tube for easy monitoring of liquid level. This industrial gauge helps ensure proper liquid fill levels in large containers up to 102 inches tall. The gauge mounts to the top of the container and requires a 1-1/4 inch NPT opening. It has a maximum temperature rating of 150°F.